Assay Details
Target Gene Details
Entrez Gene ID: | 57464 |
Gene Name: | striatin interacting protein 2 |
Gene Aliases: |
FAM40B, FAR11B |
Location: |
Chr.7:129434433-129488399 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRIP2 | NM_001134336.1 | NP_001127808.1 | ||
| NM_020704.2 | NP_065755.1 | |||
| XM_006716069.3 | XP_006716132.1 | |||
| XM_011516435.1 | XP_011514737.1 | |||
| XM_017012470.1 | XP_016867959.1 | |||
| XM_017012472.1 | XP_016867961.1 | |||
| AB032996.1 | ||||
| AK290165.1 | ||||
| BC019064.2 | AAH19064.1 | |||
| DA409007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831128 | Chr.7:129423199 - 129600959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map