Assay Details
Target Gene Details
Entrez Gene ID: | 100129460 |
Gene Name: | DPY19L1 pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.7:32580939-32719168 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPY19L1P1 | NR_036680.1 | |||
| AK026768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67223828] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map