Assay Details
Target Gene Details
Entrez Gene ID: | 57180 |
Gene Name: | ARP3 actin related protein 3 homolog B |
Gene Aliases: |
ARP11, ARP3BETA |
Location: |
Chr.7:152759749-153070979 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR3B | NM_001040135.2 | NP_001035225.1 | ||
| NM_020445.5 | NP_065178.1 | |||
| NR_073000.1 | ||||
| NR_073001.1 | ||||
| XM_006716065.2 | XP_006716128.1 | |||
| XM_011516425.2 | XP_011514727.1 | |||
| XM_011516426.2 | XP_011514728.1 | |||
| XM_017012460.1 | XP_016867949.1 | |||
| XM_017012461.1 | XP_016867950.1 | |||
| XM_017012462.1 | XP_016867951.1 | |||
| XM_017012463.1 | XP_016867952.1 | |||
| XM_017012464.1 | XP_016867953.1 | |||
| XM_017012465.1 | XP_016867954.1 | |||
| AB209174.1 | BAD92411.1 | |||
| AF023453.1 | AAC98904.1 | |||
| AF086920.1 | AAP97150.1 | |||
| AK001198.1 | ||||
| AK294292.1 | ||||
| AK315534.1 | ||||
| AK316078.1 | ||||
| BC008682.1 | AAH08682.1 | |||
| BC015207.1 | AAH15207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3615531 | Chr.7:152820301 - 152885417 on Build GRCh38 | Gain |
|
| esv3615532 | Chr.7:152837608 - 152876169 on Build GRCh38 | Gain |
|
| nsv831193 | Chr.7:152656459 - 152895593 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77537244] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map