Assay Details
Target Gene Details
Entrez Gene ID: | 2861 |
Gene Name: | G protein-coupled receptor 37 |
Gene Aliases: |
EDNRBL, PAELR, hET(B)R-LP |
Location: |
Chr.7:124745601-124766025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR37 | NM_005302.3 | NP_005293.1 | ||
| AF017262.1 | AAB70008.1 | |||
| AF502281.2 | AAM18625.2 | |||
| BC040007.1 | AAH40007.1 |
More Information
Set Membership: |
|

Genomic Map