Assay Details
Target Gene Details
Entrez Gene ID: | 392617 |
Gene Name: | extracellular leucine rich repeat and fibronectin type III domain containing 1 |
Gene Aliases: |
PPP1R28 |
Location: |
Chr.7:1666075-1747954 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELFN1 | NM_001128636.2 | NP_001122108.1 | ||
| XM_006715725.3 | XP_006715788.1 | |||
| XM_006715726.2 | XP_006715789.1 | |||
| XM_006715727.3 | XP_006715790.1 | |||
| XM_011515397.2 | XP_011513699.1 | |||
| XM_011515398.2 | XP_011513700.1 | |||
| XM_011515401.2 | XP_011513703.1 | |||
| XM_017012203.1 | XP_016867692.1 | |||
| AI962473.1 |
Target Gene Details
Entrez Gene ID: | 101927125 |
Gene Name: | ELFN1 antisense RNA 1 |
Gene Aliases: |
MYCLo-2 |
Location: |
Chr.7:1738630-1742310 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELFN1-AS1 | NR_120508.1 | |||
| NR_120509.1 | ||||
| NR_120510.1 | ||||
| HO663747.1 | ||||
| HY180852.1 | ||||
| JX046910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464248 | Chr.7:1612264 - 1802771 on Build GRCh38 | Gain |
|
| nsv950787 | Chr.7:1723165 - 1751464 on Build GRCh38 | Deletion |
|
| nsv830881 | Chr.7:1638331 - 1833504 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map