Assay Details
Target Gene Details
Entrez Gene ID: | 222068 |
Gene Name: | transmembrane p24 trafficking protein 4 |
Gene Aliases: |
ERS25, GMP25iso, HNLF, p24a3, p24alpha3 |
Location: |
Chr.7:44577894-44582295 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED4 | NM_001303059.1 | 4 | 1879 | NP_001289988.1 |
| NM_001303061.1 | 5 | 2019 | NP_001289990.1 | |
| NM_001303062.1 | 4 | 2249 | NP_001289991.1 | |
| NM_182547.3 | 5 | 2026 | NP_872353.2 |
More Information
Set Membership: |
|

Genomic Map