Assay Details
Target Gene Details
Entrez Gene ID: | 51422 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit gamma 2 |
Gene Aliases: |
AAKG, AAKG2, CMH6, H91620p, WPWS |
Location: |
Chr.7:151556114-151877231 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831189 | Chr.7:151452021 - 151622168 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map