Assay Details
Target Gene Details
Entrez Gene ID: | 5577 |
Gene Name: | protein kinase cAMP-dependent type II regulatory subunit beta |
Gene Aliases: |
PRKAR2, RII-BETA |
Location: |
Chr.7:107044728-107161811 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAR2B | NM_002736.2 | NP_002727.2 | ||
| XM_011516398.1 | XP_011514700.1 | |||
| AK128639.1 | ||||
| AK291441.1 | ||||
| AL559918.3 | ||||
| BC075800.1 | AAH75800.1 | |||
| M31158.1 | AAA60099.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831087 | Chr.7:107033020 - 107232015 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map