Assay Details
Target Gene Details
Entrez Gene ID: | 101927524 |
Gene Name: | MEOX2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.7:15688378-15696892 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEOX2-AS1 | NR_110094.1 | |||
| DA829065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830911 | Chr.7:15626682 - 15778037 on Build GRCh38 | Gain |
|
| nsv523199 | Chr.7:15542583 - 16002971 on Build GRCh38 | Gain |
|
| nsv606283 | Chr.7:14536538 - 15793555 on Build GRCh38 | Gain |
|

Genomic Map