Assay Details
Target Gene Details
Entrez Gene ID: | 63917 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 11 |
Gene Aliases: |
GALNAC-T11, GALNACT11 |
Location: |
Chr.7:152025674-152122347 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT11 | NM_001304514.1 | NP_001291443.1 | ||
| NM_022087.3 | NP_071370.2 | |||
| XM_006716082.2 | XP_006716145.1 | |||
| XM_006716083.2 | XP_006716146.1 | |||
| XM_006716084.2 | XP_006716147.1 | |||
| XM_017012499.1 | XP_016867988.1 | |||
| XM_017012500.1 | XP_016867989.1 | |||
| AK025287.1 | BAB15105.1 | |||
| AK026056.1 | BAB15338.1 | |||
| AK124934.1 | ||||
| AK296097.1 | ||||
| AK298911.1 | ||||
| Y12434.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv609045 | Chr.7:152094085 - 152201919 on Build GRCh38 | Gain |
|
| nsv1029778 | Chr.7:151966767 - 152375920 on Build GRCh38 | Gain |
|
| esv3576482 | Chr.7:152058842 - 152259368 on Build GRCh38 | Gain |
|
| nsv831191 | Chr.7:152076503 - 152260357 on Build GRCh38 | Gain |
|
| dgv6758n100 | Chr.7:152070390 - 152305071 on Build GRCh38 | Gain |
|
| nsv1020487 | Chr.7:151946824 - 152195828 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map