Assay Details
Target Gene Details
Entrez Gene ID: | 51678 |
Gene Name: | membrane palmitoylated protein 6 |
Gene Aliases: |
PALS2, VAM-1, VAM1, p55T |
Location: |
Chr.7:24573346-24694086 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP6 | NM_001303037.1 | NP_001289966.1 | ||
| NM_016447.3 | NP_057531.2 | |||
| XM_006715738.3 | XP_006715801.1 | |||
| XM_011515425.2 | XP_011513727.1 | |||
| XM_017012316.1 | XP_016867805.1 | |||
| XM_017012317.1 | XP_016867806.1 | |||
| AF162130.2 | AAD45919.2 | |||
| AL136836.1 | CAB66770.1 | |||
| BC023638.2 | AAH23638.1 | |||
| BG766138.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518495 | Chr.7:24576855 - 24591626 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142505588,rs79968383] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map