Assay Details
Target Gene Details
Entrez Gene ID: | 2913 |
Gene Name: | glutamate metabotropic receptor 3 |
Gene Aliases: |
GLUR3, GPRC1C, MGLUR3, mGlu3 |
Location: |
Chr.7:86643914-86864879 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM3 | NM_000840.2 | NP_000831.2 | ||
| XM_011516088.1 | XP_011514390.1 | |||
| XM_017012073.1 | XP_016867562.1 | |||
| AK291406.1 | ||||
| AK294178.1 | ||||
| AK294701.1 | ||||
| BC022496.1 | AAH22496.2 | |||
| BC041407.1 | AAH41407.1 | |||
| DQ315361.1 | ABC47402.1 | |||
| DQ315362.1 | ||||
| DQ315363.1 | ||||
| X77748.1 | CAA54796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831053 | Chr.7:86638855 - 86811683 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146976279] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map