Assay Details
Target Gene Details
Entrez Gene ID: | 1811 |
Gene Name: | solute carrier family 26 member 3 |
Gene Aliases: |
CLD, DRA |
Location: |
Chr.7:107765467-107803233 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A3 | NM_000111.2 | NP_000102.1 | ||
| XM_011515867.2 | XP_011514169.1 | |||
| AK300651.1 | ||||
| BC025671.1 | AAH25671.1 | |||
| BX640837.1 | CAE45910.1 | |||
| L02785.1 | AAA58443.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2556506 | Chr.7:107772359 - 107773867 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map