Assay Details
Target Gene Details
Entrez Gene ID: | 100505483 |
Gene Name: | PRKAG2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:151877042-151879223 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAG2-AS1 | NR_038926.1 | |||
| KR153483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027917 | Chr.7:151791285 - 151893200 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73470759] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map