Assay Details
Target Gene Details
Entrez Gene ID: | 375612 |
Gene Name: | lipoma HMGIC fusion partner-like 3 |
Gene Aliases: |
LHFPL4 |
Location: |
Chr.7:104328656-104908558 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL3 | NM_199000.2 | NP_945351.1 | ||
| XM_005250327.1 | XP_005250384.1 | |||
| AK124251.1 | ||||
| AK299759.1 | ||||
| AY260763.1 | AAP14954.1 | |||
| BC129937.1 | ||||
| BC129938.1 | ||||
| BC129941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608065 | Chr.7:104177149 - 104350307 on Build GRCh38 | Gain |
|
| nsv608067 | Chr.7:104303789 - 104388960 on Build GRCh38 | Loss |
|
| esv3891172 | Chr.7:103796156 - 104449969 on Build GRCh38 | Gain |
|

Genomic Map