Assay Details
Target Gene Details
Entrez Gene ID: | 51666 |
Gene Name: | ankyrin repeat and SOCS box containing 4 |
Gene Aliases: |
ASB-4 |
Location: |
Chr.7:95485439-95544738 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB4 | NM_016116.2 | NP_057200.1 | ||
| NM_145872.2 | NP_665879.1 | |||
| XM_017012303.1 | XP_016867792.1 | |||
| AF156779.1 | AAD41896.1 | |||
| BC025024.1 | AAH25024.1 | |||
| BC113479.1 | ||||
| BC113481.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map