Assay Details
Target Gene Details
Entrez Gene ID: | 729920 |
Gene Name: | isoprenoid synthase domain containing |
Gene Aliases: |
MDDGA7, MDDGC7, Nip, hCG_1745121 |
Location: |
Chr.7:16087527-16422562 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISPD | NM_001101417.3 | 9 | 4007 | NP_001094887.1 |
| NM_001101426.3 | 10 | 4157 | NP_001094896.1 | |
| AK124805.1 | 1 | 757 | BAC85954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734048 | Chr.7:16088848 - 16089210 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map