Assay Details
Target Gene Details
Entrez Gene ID: | 5575 |
Gene Name: | protein kinase cAMP-dependent type I regulatory subunit beta |
Gene Aliases: |
PRKAR1 |
Location: |
Chr.7:549185-727676 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAR1B | NM_001164758.1 | NP_001158230.1 | ||
| NM_001164759.1 | NP_001158231.1 | |||
| NM_001164760.1 | NP_001158232.1 | |||
| NM_001164761.1 | NP_001158233.1 | |||
| NM_001164762.1 | NP_001158234.1 | |||
| NM_002735.2 | NP_002726.1 | |||
| XM_011515446.2 | XP_011513748.1 | |||
| AK098822.1 | ||||
| AK289464.1 | ||||
| AK297337.1 | ||||
| AK315951.1 | ||||
| AK315990.1 | ||||
| AL833563.1 | ||||
| BC026734.1 | AAH26734.1 | |||
| BC036828.1 | AAH36828.2 | |||
| CT003793.1 | ||||
| DA210679.1 | ||||
| M65066.1 | AAC37564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3731e59 | Chr.7:653289 - 655287 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map