Assay Details
Target Gene Details
Entrez Gene ID: | 5879 |
Gene Name: | ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1) |
Gene Aliases: |
MIG5, Rac-1, TC-25, p21-Rac1 |
Location: |
Chr.7:6374495-6403967 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAC1 | NM_006908.4 | NP_008839.2 | ||
| NM_018890.3 | NP_061485.1 | |||
| AB209410.1 | BAD92647.1 | |||
| AB451238.1 | ||||
| AB451363.1 | ||||
| AF136373.1 | AAD30547.1 | |||
| AF498964.1 | AAM21111.1 | |||
| AJ132694.1 | CAA10732.1 | |||
| AK054993.1 | ||||
| AK291806.1 | ||||
| AK291815.1 | ||||
| AY279384.1 | AAQ16632.1 | |||
| BC004247.1 | AAH04247.1 | |||
| BC050687.1 | AAH50687.1 | |||
| BC107748.1 | AAI07749.1 | |||
| BG437715.1 | ||||
| BQ689587.1 | ||||
| BT007121.1 | AAP35785.1 | |||
| M29870.1 | AAA36537.1 | |||
| M31467.1 | AAA36544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020236 | Chr.7:6209299 - 6650746 on Build GRCh38 | Gain |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv956982 | Chr.7:6352597 - 6376151 on Build GRCh38 | Deletion |
|
| nsv605999 | Chr.7:6357088 - 6433250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map