Assay Details
Target Gene Details
Entrez Gene ID: | 6717 |
Gene Name: | sorcin |
Gene Aliases: |
CP-22, CP22, SCN, V19 |
Location: |
Chr.7:88205115-88226993 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRI | NM_001256891.1 | NP_001243820.1 | ||
| NM_001256892.1 | NP_001243821.1 | |||
| NM_003130.3 | NP_003121.1 | |||
| NM_198901.1 | NP_944490.1 | |||
| XM_011516528.1 | XP_011514830.1 | |||
| AK129770.1 | ||||
| AK295225.1 | ||||
| AK296601.1 | ||||
| AK310366.1 | ||||
| AL039198.1 | ||||
| BC011025.1 | AAH11025.1 | |||
| BI524104.1 | ||||
| BI559592.1 | ||||
| BI597920.1 | ||||
| BM739144.1 | ||||
| DA268050.1 | ||||
| L12387.1 | AAA92155.1 | |||
| M32886.1 | AAA60588.1 |
More Information
Set Membership: |
|

Genomic Map