Assay Details
Target Gene Details
Entrez Gene ID: | 10466 |
Gene Name: | component of oligomeric golgi complex 5 |
Gene Aliases: |
CDG2I, GOLTC1, GTC90 |
Location: |
Chr.7:107201743-107564514 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG5 | NM_001161520.1 | NP_001154992.1 | ||
| NM_006348.3 | NP_006339.3 | |||
| NM_181733.2 | NP_859422.2 | |||
| AF058718.1 | AAC69276.1 | |||
| AK314270.1 | ||||
| BC068540.1 | AAH68540.1 | |||
| BI597200.1 |
Target Gene Details
Entrez Gene ID: | 2845 |
Gene Name: | G protein-coupled receptor 22 |
Gene Aliases: |
- |
Location: |
Chr.7:107470057-107476207 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR22 | XM_011516056.2 | 2 | 2773 | XP_011514358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024793 | Chr.7:107260432 - 107529867 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map