Assay Details
Target Gene Details
Entrez Gene ID: | 1395 |
Gene Name: | corticotropin releasing hormone receptor 2 |
Gene Aliases: |
CRF-RB, CRF2, CRFR2, HM-CRF |
Location: |
Chr.7:30651844-30700103 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRHR2 | NM_001202475.1 | NP_001189404.1 | ||
| NM_001202481.1 | NP_001189410.1 | |||
| NM_001202482.1 | NP_001189411.1 | |||
| NM_001202483.1 | NP_001189412.1 | |||
| NM_001883.4 | NP_001874.2 | |||
| XM_011515127.2 | XP_011513429.1 | |||
| XM_011515128.2 | XP_011513430.1 | |||
| XM_011515129.2 | XP_011513431.1 | |||
| XM_017011751.1 | XP_016867240.1 | |||
| XM_017011752.1 | XP_016867241.1 | |||
| AF011406.1 | AAB94503.1 | |||
| AF019381.1 | AAB94562.1 | |||
| AK313661.1 | ||||
| AY449734.1 | AAR18078.1 | |||
| BC096830.1 | AAH96830.1 | |||
| EU012439.1 | ||||
| EU012440.1 | ||||
| EU012441.1 | ||||
| EU012442.1 | ||||
| EU012443.1 | ||||
| FJ200484.1 | ||||
| U34587.1 | AAA91320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6316n100 | Chr.7:30609596 - 31145482 on Build GRCh38 | Gain |
|
| nsv830938 | Chr.7:30644614 - 30857907 on Build GRCh38 | Gain |
|
| nsv527038 | Chr.7:30617851 - 31122331 on Build GRCh38 | Gain |
|
| esv2761316 | Chr.7:30615466 - 31127062 on Build GRCh38 | Gain |
|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map