Assay Details
Target Gene Details
Entrez Gene ID: | 889 |
Gene Name: | KRIT1, ankyrin repeat containing |
Gene Aliases: |
CAM, CCM1 |
Location: |
Chr.7:92198969-92246128 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
More Information
Set Membership: |
|

Genomic Map