Assay Details
Target Gene Details
Entrez Gene ID: | 57541 |
Gene Name: | zinc finger protein 398 |
Gene Aliases: |
P51, P71, ZER6 |
Location: |
Chr.7:149126416-149183042 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF398 | NM_020781.3 | NP_065832.1 | ||
| NM_170686.2 | NP_733787.1 | |||
| XM_011516439.2 | XP_011514741.1 | |||
| XM_011516440.2 | XP_011514742.1 | |||
| XM_011516441.2 | XP_011514743.1 | |||
| AK026380.1 | ||||
| AK290499.1 | ||||
| AK301890.1 | ||||
| AY049743.1 | AAK92788.1 | |||
| AY049744.1 | AAK92789.1 | |||
| BC043295.2 | AAH43295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018754 | Chr.7:149076805 - 149173707 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map