Assay Details
Target Gene Details
Entrez Gene ID: | 6742 |
Gene Name: | single stranded DNA binding protein 1 |
Gene Aliases: |
Mt-SSB, SOSS-B1, SSBP, mtSSB |
Location: |
Chr.7:141738309-141750488 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSBP1 | NM_001256510.1 | NP_001243439.1 | ||
| NM_001256511.1 | NP_001243440.1 | |||
| NM_001256512.1 | NP_001243441.1 | |||
| NM_001256513.1 | NP_001243442.1 | |||
| NM_003143.2 | NP_003134.1 | |||
| NR_046269.1 | ||||
| AF277319.1 | AAK69112.1 | |||
| AK313033.1 | ||||
| BC000895.1 | AAH00895.1 | |||
| BC093054.1 | AAH93054.1 | |||
| BF131408.1 | ||||
| BP308841.1 | ||||
| BQ959702.1 | ||||
| BU532904.1 | ||||
| M94556.1 | AAA36332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34135 | Chr.7:141347654 - 141793098 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113469479,rs142524008] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map