Assay Details
Target Gene Details
Entrez Gene ID: | 723809 |
Gene Name: | LHFPL3 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.7:104894628-104926645 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL3-AS2 | NR_027374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436902 | Chr.7:104802439 - 105205347 on Build GRCh38 | Insertion |
|
| nsv831086 | Chr.7:104782056 - 104956319 on Build GRCh38 | Loss |
|

Genomic Map