Assay Details
Target Gene Details
Entrez Gene ID: | 22798 |
Gene Name: | laminin subunit beta 4 |
Gene Aliases: |
- |
Location: |
Chr.7:108011662-108130356 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMB4 | NM_001318046.1 | NP_001304975.1 | ||
| NM_007356.2 | NP_031382.2 | |||
| XM_011515975.1 | XP_011514277.1 | |||
| XM_011515978.1 | XP_011514280.1 | |||
| XM_011515979.2 | XP_011514281.1 | |||
| XM_011515980.2 | XP_011514282.1 | |||
| XM_017011879.1 | XP_016867368.1 | |||
| XM_017011880.1 | XP_016867369.1 | |||
| AF028816.1 | AAC95123.1 | |||
| AK301777.1 | ||||
| BC045172.2 | AAH45172.2 | |||
| BC140804.1 | ||||
| BC144591.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831088 | Chr.7:107856655 - 108053235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map