Assay Details
Target Gene Details
Entrez Gene ID: | 7982 |
Gene Name: | suppression of tumorigenicity 7 |
Gene Aliases: |
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7 |
Location: |
Chr.7:116953327-117230023 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST7 | NM_018412.3 | NP_060882.2 | ||
| NM_021908.2 | NP_068708.1 | |||
| AF234882.1 | AAF85945.1 | |||
| AF234883.1 | AAF85946.1 | |||
| AK292372.1 | ||||
| AK298540.1 | ||||
| BC030954.1 | AAH30954.1 | |||
| BC075855.1 | AAH75855.1 | |||
| KJ534947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1324e214 | Chr.7:116905405 - 117056329 on Build GRCh38 | Gain |
|
| esv3891196 | Chr.7:116905706 - 117032710 on Build GRCh38 | Gain |
|
| nsv527321 | Chr.7:116994994 - 117020749 on Build GRCh38 | Loss |
|
| esv3614704 | Chr.7:116947077 - 117046223 on Build GRCh38 | Gain |
|
| nsv831107 | Chr.7:116774680 - 117009412 on Build GRCh38 | Gain |
|

Genomic Map