Assay Details
Target Gene Details
Entrez Gene ID: | 10392 |
Gene Name: | nucleotide binding oligomerization domain containing 1 |
Gene Aliases: |
CARD4, CLR7.1, NLRC1 |
Location: |
Chr.7:30424527-30478840 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOD1 | NM_006092.2 | NP_006083.1 | ||
| XM_005249568.1 | XP_005249625.1 | |||
| XM_005249572.1 | XP_005249629.1 | |||
| XM_005249576.1 | XP_005249633.1 | |||
| XM_006715633.2 | XP_006715696.1 | |||
| XM_011515079.1 | XP_011513381.1 | |||
| XM_011515080.2 | XP_011513382.1 | |||
| XM_011515081.2 | XP_011513383.1 | |||
| XM_011515083.1 | XP_011513385.1 | |||
| XM_011515084.1 | XP_011513386.1 | |||
| XM_011515085.1 | XP_011513387.1 | |||
| XM_017011674.1 | XP_016867163.1 | |||
| XM_017011675.1 | XP_016867164.1 | |||
| AF113925.1 | AAD28350.1 | |||
| AF126484.1 | AAD29125.1 | |||
| AK023969.1 | ||||
| BC040339.1 | AAH40339.1 | |||
| KU178514.1 | ||||
| KU178515.1 | ||||
| KU178516.1 | ||||
| KU178517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map