Assay Details
Target Gene Details
Entrez Gene ID: | 401409 |
Gene Name: | RAB19, member RAS oncogene family |
Gene Aliases: |
RAB19B |
Location: |
Chr.7:140403740-140426881 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB19 | NM_001008749.2 | NP_001008749.2 | ||
| XM_005249996.3 | XP_005250053.1 | |||
| XM_005249997.3 | XP_005250054.1 | |||
| XM_005249998.3 | XP_005250055.1 | |||
| XM_017012207.1 | 1 | 93 | XP_016867696.1 | |
| DN996121.1 | ||||
| DY655778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762706 | Chr.7:140354370 - 140419829 on Build GRCh38 | Loss |
|
| nsv522190 | Chr.7:140366611 - 140430863 on Build GRCh38 | Gain |
|
| nsv1022919 | Chr.7:140343499 - 140420746 on Build GRCh38 | Gain |
|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map