Assay Details
Target Gene Details
Entrez Gene ID: | 1687 |
Gene Name: | DFNA5, deafness associated tumor suppressor |
Gene Aliases: |
ICERE-1 |
Location: |
Chr.7:24698355-24758020 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DFNA5 | NM_001127453.1 | NP_001120925.1 | ||
| NM_001127454.1 | NP_001120926.1 | |||
| NM_004403.2 | NP_004394.1 | |||
| XM_017011802.1 | XP_016867291.1 | |||
| AF007790.2 | AAC39635.2 | |||
| AF073308.1 | AAC69324.1 | |||
| AK094714.1 | ||||
| AK290775.1 | ||||
| AK314402.1 | ||||
| BC099911.1 | ||||
| BC125065.1 | ||||
| BC125066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891089 | Chr.7:24704690 - 24729781 on Build GRCh38 | Loss |
|
| nsv520276 | Chr.7:24649588 - 25027423 on Build GRCh38 | Gain |
|
| nsv527599 | Chr.7:24591626 - 24708291 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map