Assay Details
Target Gene Details
Entrez Gene ID: | 1129 |
Gene Name: | cholinergic receptor muscarinic 2 |
Gene Aliases: |
HM2 |
Location: |
Chr.7:136868652-137020255 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRM2 | NM_000739.2 | NP_000730.1 | ||
| NM_001006626.1 | NP_001006627.1 | |||
| NM_001006627.1 | NP_001006628.1 | |||
| NM_001006628.1 | NP_001006629.1 | |||
| NM_001006629.1 | NP_001006630.1 | |||
| NM_001006630.1 | NP_001006631.1 | |||
| NM_001006631.1 | NP_001006632.1 | |||
| NM_001006632.1 | NP_001006633.1 | |||
| AK290236.1 | ||||
| AL832585.1 | ||||
| AY034603.2 | AAO23567.1 | |||
| AY034604.2 | AAO23568.1 | |||
| AY034605.2 | AAO23569.1 | |||
| AY219703.1 | AAO65595.1 | |||
| AY219704.1 | AAO65596.1 | |||
| AY219705.1 | AAO65597.1 | |||
| AY456125.1 | AAR14890.1 | |||
| AY456126.1 | AAR14891.1 | |||
| AY456127.1 | AAR14892.1 |
Target Gene Details
Entrez Gene ID: | 349160 |
Gene Name: | uncharacterized LOC349160 |
Gene Aliases: |
- |
Location: |
Chr.7:136898773-137164341 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC349160 | NR_046103.1 | |||
| AF279773.1 | ||||
| AK097470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831147 | Chr.7:136898697 - 137070994 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138090564] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map