Assay Details
Target Gene Details
Entrez Gene ID: | 168455 |
Gene Name: | coiled-coil domain containing 71-like |
Gene Aliases: |
C7orf74 |
Location: |
Chr.7:106656765-106661188 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC71L | NM_175884.4 | 1 | 3989 | NP_787080.2 |
| AI674055.1 | 1 | 438 | ||
| AK021498.1 | 1 | 1046 | ||
| AK098422.1 | 1 | 1758 |

Genomic Map