Assay Details
Target Gene Details
Entrez Gene ID: | 51251 |
Gene Name: | 5'-nucleotidase, cytosolic IIIA |
Gene Aliases: |
NT5C3, P5'N-1, P5N-1, PN-I, POMP, PSN1, UMPH, UMPH1, cN-III, hUMP1, p36 |
Location: |
Chr.7:33014113-33062797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5C3A | NM_001002009.2 | NP_001002009.1 | ||
| NM_001002010.2 | NP_001002010.1 | |||
| AF151067.1 | AAF36153.1 | |||
| AF312735.1 | AAG33630.1 | |||
| AK290118.1 | ||||
| AK314109.1 | ||||
| AL136716.1 | CAB66650.1 | |||
| BC013292.2 | AAH13292.2 | |||
| BC066914.1 | AAH66914.1 | |||
| BC071652.1 | ||||
| CB127255.1 | ||||
| CB137440.1 | ||||
| CR533518.1 | CAG38549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029220 | Chr.7:32881872 - 33222578 on Build GRCh38 | Gain |
|
| dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion |
|
| nsv830942 | Chr.7:33008463 - 33207123 on Build GRCh38 | Gain |
|
| esv2734229 | Chr.7:32916268 - 33093711 on Build GRCh38 | Deletion |
|
| nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion |
|
| nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72555720] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map