Assay Details
Target Gene Details
Entrez Gene ID: | 100289187 |
Gene Name: | transmembrane protein 225-like |
Gene Aliases: |
- |
Location: |
Chr.7:99597936-99610833 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GS1-259H13.2 | NM_001195541.1 | NP_001182470.1 | ||
| NM_001195542.1 | NP_001182471.1 | |||
| NM_001195543.1 | NP_001182472.1 | |||
| XM_005250088.4 | XP_005250145.1 | |||
| XM_011515691.2 | XP_011513993.1 | |||
| XM_011515692.2 | XP_011513994.1 | |||
| XM_011515694.2 | XP_011513996.1 | |||
| XM_011515695.2 | XP_011513997.1 | |||
| XM_011515696.2 | XP_011513998.1 | |||
| XM_011515697.1 | XP_011513999.1 | |||
| XM_011515699.2 | XP_011514001.1 | |||
| XM_011515700.2 | XP_011514002.1 | |||
| XM_017011629.1 | XP_016867118.1 | |||
| BI559535.1 | ||||
| DB054606.1 | ||||
| DB442398.1 | ||||
| DB456868.1 | ||||
| DB462723.1 |

Genomic Map