Assay Details
Target Gene Details
Entrez Gene ID: | 816 |
Gene Name: | calcium/calmodulin dependent protein kinase II beta |
Gene Aliases: |
CAM2, CAMK2, CAMKB |
Location: |
Chr.7:44217150-44325994 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK2B | NM_001220.4 | NP_001211.3 | ||
| NM_001293170.1 | NP_001280099.1 | |||
| NM_172078.2 | NP_742075.1 | |||
| NM_172079.2 | NP_742076.1 | |||
| NM_172080.2 | NP_742077.1 | |||
| NM_172081.2 | NP_742078.1 | |||
| NM_172082.2 | NP_742079.1 | |||
| NM_172083.2 | NP_742080.1 | |||
| NM_172084.2 | NP_742081.1 | |||
| AB073352.1 | ||||
| AK290148.1 | ||||
| BQ130578.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606751 | Chr.7:44213422 - 44232814 on Build GRCh38 | Gain |
|
| nsv509195 | Chr.7:44147118 - 44261782 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map