Assay Details
Target Gene Details
Entrez Gene ID: | 105375166 |
Gene Name: | uncharacterized LOC105375166 |
Gene Aliases: |
- |
Location: |
Chr.7:15667947-15681980 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375166 | NR_134258.1 | |||
| BX538274.1 |
Target Gene Details
Entrez Gene ID: | 4223 |
Gene Name: | mesenchyme homeobox 2 |
Gene Aliases: |
GAX, MOX2 |
Location: |
Chr.7:15611212-15686683 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEOX2 | NM_005924.4 | NP_005915.2 | ||
| AK309472.1 | ||||
| AK313386.1 | ||||
| BC017021.1 | AAH17021.1 | |||
| CR536553.1 | CAG38790.1 | |||
| L36328.1 | AAA58497.1 | |||
| X82629.1 | CAA57949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830911 | Chr.7:15626682 - 15778037 on Build GRCh38 | Gain |
|
| nsv523199 | Chr.7:15542583 - 16002971 on Build GRCh38 | Gain |
|
| nsv606283 | Chr.7:14536538 - 15793555 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map