Assay Details
Target Gene Details
Entrez Gene ID: | 4852 |
Gene Name: | neuropeptide Y |
Gene Aliases: |
PYY4 |
Location: |
Chr.7:24284188-24291865 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPY | NM_000905.3 | NP_000896.1 | ||
| AK312156.1 | ||||
| BC029497.1 | AAH29497.1 | |||
| K01911.1 | AAA59944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2673027 | Chr.7:23679603 - 24432457 on Build GRCh38 | Deletion |
|
| nsv830927 | Chr.7:24262646 - 24438857 on Build GRCh38 | Gain |
|
| nsv517053 | Chr.7:24224894 - 24386936 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map