Assay Details
Target Gene Details
Entrez Gene ID: | 9844 |
Gene Name: | engulfment and cell motility 1 |
Gene Aliases: |
CED-12, CED12, ELMO-1 |
Location: |
Chr.7:36852906-37449409 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMO1 | NM_001039459.2 | 7 | 2614 | NP_001034548.1 |
| NM_001206480.2 | 22 | 4393 | NP_001193409.1 | |
| NM_001206482.1 | 22 | 4563 | NP_001193411.1 | |
| NM_014800.10 | 22 | 4204 | NP_055615.8 | |
| NM_130442.3 | 7 | 2760 | NP_569709.1 | |
| NR_038120.1 | 6 | 2647 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5701 | Chr.7:36844883 - 36862375 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map