Assay Details
Target Gene Details
Entrez Gene ID: | 285973 |
Gene Name: | autophagy related 9B |
Gene Aliases: |
APG9L2, NOS3AS, SONE |
Location: |
Chr.7:151012209-151024499 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG9B | NM_001317056.1 | NP_001303985.1 | ||
| NR_073169.1 | ||||
| NR_133652.1 | ||||
| XM_011516065.1 | XP_011514367.1 | |||
| XM_011516066.2 | XP_011514368.1 | |||
| AK096734.1 | BAC04853.1 | |||
| AY316116.1 | AAQ86941.1 | |||
| AY515311.1 | AAS87212.1 | |||
| BC128587.1 | ||||
| BX420361.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526410 | Chr.7:151017725 - 151078219 on Build GRCh38 | Loss |
|
| nsv951648 | Chr.7:150931413 - 151106413 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map