Assay Details
Target Gene Details
Entrez Gene ID: | 6916 |
Gene Name: | thromboxane A synthase 1 |
Gene Aliases: |
BDPLT14, CYP5, CYP5A1, GHOSAL, THAS, TS, TXAS, TXS |
Location: |
Chr.7:139778248-140020325 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBXAS1 | NM_001130966.2 | NP_001124438.1 | ||
| NM_001166254.1 | NP_001159726.1 | |||
| AK223466.1 | BAD97186.1 | |||
| BC014117.2 | AAH14117.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464732 | Chr.7:139724223 - 139954457 on Build GRCh38 | Gain |
|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
| dgv6641n100 | Chr.7:139693076 - 139984244 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73735623] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map