Assay Details
Target Gene Details
Entrez Gene ID: | 7988 |
Gene Name: | zinc finger protein 212 |
Gene Aliases: |
C2H2-150, ZNF182, ZNFC150 |
Location: |
Chr.7:149239648-149255609 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF212 | NM_012256.3 | NP_036388.2 | ||
| XM_005250055.2 | XP_005250112.2 | |||
| XM_011516581.2 | XP_011514883.2 | |||
| XM_011516582.1 | XP_011514884.1 | |||
| XM_017012646.1 | XP_016868135.1 | |||
| XM_017012647.1 | XP_016868136.1 | |||
| XM_017012648.1 | XP_016868137.1 | |||
| AK074821.1 | ||||
| AK315086.1 | ||||
| BC022785.2 | AAH22785.2 | |||
| U38864.1 | AAC51296.1 |
More Information
Set Membership: |
|

Genomic Map