Assay Details
Target Gene Details
Entrez Gene ID: | 401337 |
Gene Name: | long intergenic non-protein coding RNA 1446 |
Gene Aliases: |
GS1-179L18.1 |
Location: |
Chr.7:53655509-53811931 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01446 | NR_038371.1 | |||
| AK127870.1 | BAC87170.1 |

Genomic Map