Assay Details
Target Gene Details
Entrez Gene ID: | 392617 |
Gene Name: | extracellular leucine rich repeat and fibronectin type III domain containing 1 |
Gene Aliases: |
PPP1R28 |
Location: |
Chr.7:1666075-1747954 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELFN1 | XM_006715725.3 | XP_006715788.1 | ||
| XM_006715726.2 | XP_006715789.1 | |||
| XM_011515397.2 | XP_011513699.1 | |||
| XM_011515398.2 | XP_011513700.1 | |||
| XM_017012203.1 | XP_016867692.1 | |||
| AI962473.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3611817 | Chr.7:1704891 - 1713240 on Build GRCh38 | Loss |
|
| nsv464248 | Chr.7:1612264 - 1802771 on Build GRCh38 | Gain |
|
| nsv830881 | Chr.7:1638331 - 1833504 on Build GRCh38 | Loss |
|
| nsv605826 | Chr.7:1640529 - 1717600 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map