Assay Details
Target Gene Details
Entrez Gene ID: | 7982 |
Gene Name: | suppression of tumorigenicity 7 |
Gene Aliases: |
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7 |
Location: |
Chr.7:116953327-117230023 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST7 | NM_018412.3 | NP_060882.2 | ||
| NM_021908.2 | NP_068708.1 | |||
| AF234882.1 | AAF85945.1 | |||
| AF234883.1 | AAF85946.1 | |||
| AJ277291.1 | CAB86869.1 | |||
| AK093212.1 | BAC04097.1 | |||
| AK292372.1 | ||||
| AK297510.1 | ||||
| AK297861.1 | ||||
| AK298540.1 | ||||
| AY009152.1 | AAG37720.1 | |||
| AY009153.1 | AAG37721.1 | |||
| BC030954.1 | AAH30954.1 | |||
| BC075855.1 | AAH75855.1 | |||
| CR936656.1 | CAI56794.1 | |||
| KJ534947.1 |
Target Gene Details
Entrez Gene ID: | 93655 |
Gene Name: | ST7 overlapping transcript 3 |
Gene Aliases: |
NCRNA00026, ST7, ST7OT3 |
Location: |
Chr.7:117182681-117209937 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST7-OT3 | NR_002332.2 | |||
| AF400044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831108 | Chr.7:117109480 - 117311957 on Build GRCh38 | Gain |
|
| nsv5916 | Chr.7:117185167 - 117230516 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116327736] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map