Assay Details
Target Gene Details
Entrez Gene ID: | 1180 |
Gene Name: | chloride voltage-gated channel 1 |
Gene Aliases: |
CLC1 |
Location: |
Chr.7:143316126-143352008 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCN1 | NM_000083.2 | NP_000074.2 | ||
| NR_046453.1 | ||||
| BC112156.1 | AAI12157.1 | |||
| BC113495.1 | ||||
| Z25884.1 | CAA81103.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831169 | Chr.7:143188183 - 143348025 on Build GRCh38 | Loss |
|
| nsv5984 | Chr.7:143305004 - 143350170 on Build GRCh38 | Deletion |
|
| nsv5985 | Chr.7:143308794 - 143338476 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map