Assay Details
Target Gene Details
Entrez Gene ID: | 57001 |
Gene Name: | succinate dehydrogenase complex assembly factor 3 |
Gene Aliases: |
ACN9, DC11, LYRM10, Sdh7 |
Location: |
Chr.7:97116593-97181763 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHAF3 | NM_020186.2 | NP_064571.1 | ||
| AF201933.1 | AAF86869.1 | |||
| BC020621.1 | AAH20621.1 | |||
| BC022858.1 | AAH22858.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79171796] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map