Assay Details
Target Gene Details
Entrez Gene ID: | 401337 |
Gene Name: | long intergenic non-protein coding RNA 1446 |
Gene Aliases: |
GS1-179L18.1 |
Location: |
Chr.7:53655509-53811931 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01446 | NR_038371.1 | |||
| AK127870.1 | BAC87170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022136 | Chr.7:53686430 - 53709528 on Build GRCh38 | Gain |
|

Genomic Map