Assay Details
Target Gene Details
Entrez Gene ID: | 54504 |
Gene Name: | carboxypeptidase, vitellogenic like |
Gene Aliases: |
HVLP |
Location: |
Chr.7:28995630-29146537 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPVL | NM_019029.2 | NP_061902.2 | ||
| NM_031311.3 | NP_112601.3 | |||
| XM_011515437.1 | XP_011513739.1 | |||
| XM_017012366.1 | XP_016867855.1 | |||
| AF106704.2 | AAG37991.2 | |||
| AF217508.1 | AAF67619.1 | |||
| AF282617.1 | AAG14348.1 | |||
| AK075433.1 | BAC11618.1 | |||
| AK124472.1 | ||||
| AK225545.1 | ||||
| AY358549.1 | AAQ88913.1 | |||
| BC016838.2 | AAH16838.1 |
Target Gene Details
Entrez Gene ID: | 100506497 |
Gene Name: | uncharacterized LOC100506497 |
Gene Aliases: |
- |
Location: |
Chr.7:28979967-29013369 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506497 | NR_038965.1 | |||
| AK124888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612679 | Chr.7:28970964 - 29086873 on Build GRCh38 | Loss |
|
| esv3612685 | Chr.7:28997136 - 29209454 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76335499] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map