Assay Details
Target Gene Details
Entrez Gene ID: | 22798 |
Gene Name: | laminin subunit beta 4 |
Gene Aliases: |
- |
Location: |
Chr.7:108011662-108130356 on Build GRCh38 |
Assay Gene Location: | Within Intron 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMB4 | XM_011515975.1 | XP_011514277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831088 | Chr.7:107856655 - 108053235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map